missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Tuner™ DE3 pLysS Competent Cells

Catalog No. 706243
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
706243 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 706243 Supplier MilliporeSigma™ Supplier No. 706243

Tuner host strains are lacZY deletion mutants of BL21, which enable adjustable levels of protein expression throughout all cells in a culture. T7 lysozyme expression suppresses basal T7 expression.

Tuner™ strains are lacZY deletion mutants of BL21 and enable adjustable levels of protein expression throughout all cells in a culture. The lac permease (lacY) mutation allows uniform entry of IPTG into all cells in the population, which produces a concentration-dependent, homogeneous level of induction. By adjusting the concentration of IPTG, expression can be regulated from very low levels up to the robust, fully induced levels commonly associated with pET vectors. Lower-level expression may enhance the solubility and activity of difficult target proteins. DE3 indicates that the host is a lysogen of ™DE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 0.4 mL