missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Rosetta-gami™ 2 (DE3) pLysS Competent Cells

Catalog No. 713524
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
713524 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 713524 Supplier MilliporeSigma™ Supplier No. 713524

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rosetta-Gami 2 host strains allows for enhanced disulfide bond formation and enhanced expression of eukaryotic proteins. T7 lysozyme expression suppresses basal T7 expression.

Rosetta-gami 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli. These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 mL