missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Rosetta™ (DE3) Competent Cells

Catalog No. 709544
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
709544 1 mL
709543 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 709544 Supplier MilliporeSigma™ Supplier No. 709544

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.

Rosetta™ host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, GGA codons on a compatible chloramphenicol-resistant plasmid. Thus the Rosetta strains provide for universal translation which is otherwise limited by the codon usage of E. coli. The tRNA genes are driven by their native promoters. In Rosetta(DE3)pLysS, the rare tRNA genes are present on the same plasmids that carries the T7 lysozyme gene. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

Specifications

Content And Storage ≤-70°C
Quantity 1 mL