missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Rosetta™ 2 (DE3) Competent Cells

Catalog No. 713973
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
713973 0.4 mL
713974 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 713973 Supplier MilliporeSigma™ Supplier No. 713973

Novagen's Rosetta™ 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. Coli.

Rosetta™ 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for 7 rare codones (AGA, AGG, AUA, CUA, GGA, CCC, and CGG) on a compatible chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 0.4 mL