Learn More
MilliporeSigma™ Novagen™ NovaBlue GigaSingles™ Competent Cells
NovaBlue GigaSingles™ Competent Cells produce greater than 1 x 10â¹ colonies/μg plasmid DNA for cloning applications requiring high-efficiency transformations.
$764.43 - $764.43
Specifications
| Efficiency | Guaranteed efficiency: >1 x 109 cfu/μg |
|---|---|
| Content And Storage | ≤-70°C |
Description
NovaBlue competent cells are for routine molecular cloning applications. NovaBlue is a K-12 strain derivative that offers high transformation efficiency, facilitates plasmid stability, and allows blue/white screening with appropriate plasmids.Specifications
| Guaranteed efficiency: >1 x 109 cfu/μg | |
| ≤-70°C |