Learn More
MilliporeSigma™ Novagen™ BL21 (DE3) pLysS Singles™ Competent Cells
BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases.
$281.02 - $484.60
Specifications
Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
Content And Storage ≤-70°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
702363
View Documents
MilliporeSigma™
702363
11 Reactions
Each for $281.02
Only null left
702364
View Documents
MilliporeSigma™
702364
22 Reactions
Each for $484.60
Only null left
Description
Description
BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Specifications
Specifications
Guaranteed efficiency: >2 x 107 cfu/μg
≤-70°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'