missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ BL21(DE3) pLysS Competent Cells

Catalog No. 694513
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
694513 0.4 mL
694514 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 694513 Supplier MilliporeSigma™ Supplier No. 694513

BL21 host strain that expresses T7 RNA polymerase and also encode T7 lysozyme that suppresses basal expression of toxic target proteins prior to induction.

BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.

Specifications

Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
Content And Storage ≤-70°C
Quantity 0.4 mL