Learn More
MilliporeSigma™ Novagen™ BL21(DE3) Competent Cells
Catalog No. 694503
Change view
Click to view available options
Quantity:
0.4 mL (2 x 0.2 mL aliquots)
1 mL (5 x 0.2 mL aliquots)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
694503 0.4 mL (2 x 0.2 mL aliquots)
694504 1 mL (5 x 0.2 mL aliquots)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 694503 Supplier MilliporeSigma™ Supplier No. 694503
Achieve greater efficiency, reproducibility and convenience with Novagen prepared competent cells. BL21(DE3) is a chemically competent E. coli cell suitable for transformation and high-level protein expression using a T7 RNA polymerase-IPTG induction system.
  • Deficient in Ion and ompT proteases, BL21 and its derivatives are ideal for many applications
    T7 expression strain
  • Lysogen of bacteriophage DE3
  • Carries copy of T7 RNA polymerase gene under control of the lacUV5 promoter
  • Suitable for production of protein from target genes cloned in appropriate T7 expression vectors, using IPTG as an inducer
Specifications
Efficiency Guaranteed efficiency: >2 x 107 cfu/μg
Content And Storage Store at ≤-70deg;C, Avoid repeated freeze/thaw
Quantity 0.4 mL (2 x 0.2 mL aliquots)