missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ AP Detection Reagent Kit
Description
- The AP Detection Reagent Kit is designed for colorimetric detection applications.
- Format: Includes sufficient NBT, BCIP and 20xAP Buffer to perform 25 blots
- Detection Method:Colorimetric.
Specifications
Specifications
| Quantity | Each |
| For Use With (Application) | For sensitive chromogenic detection of alkaline phosphatase conjugates |
| Product Type | AP Detection Reagent Kit |