missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Ndiff™ Chemicon™ Neuro-2 Medium Supplement (200x)

Catalog No. SCM012
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM012 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM012 Supplier MilliporeSigma™ Supplier No. SCM012

a serum-free, N2-like supplement for the in vitro differentiation of murine embryonic stem (ES) cells into post-mitotic neurons particularly via monolayer differentiation.

Originally developed in Professor Austin Smith's laboratory, NDiff™ Neuro-2 Medium Supplement is a serum-free, N2-like supplement for the

Specifications

Content And Storage -20°C
Serum Level Serum-free
For Use With (Application) Stem cell culture and cell culture
Quantity 5 mL