Learn More
MilliporeSigma™ N21 Medium Supplement (50x)
Catalog No. SCM081
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
SCM081 10 mL
1 options
Catalog No. SCM081 Supplier MilliporeSigma™ Supplier No. SCM081
Only null left
an optimized serum-free supplement developed for isolation and expansion of neurons from mouse and rat hippocampus.
N21 Medium Supplement is an optimized serum-free supplement developed for isolation and expansion of neurons from mouse and rat hippocampus. In addition to its use for long term neuronal survival, it has also been validated for serum-free culture of mouse ES cells and human ES cell derived neural progenitor cells.
Specifications
For Use With (Application) Stem cell culture
Quantity 10 mL