Learn More
MilliporeSigma™ MultiScreen™ Vacuum Manifold Basic Kit
Catalog No. MAVM0960R
Change view
Click to view available options
Vacuum manifold basic kit
Catalog No.
MAVM0960R Vacuum manifold basic kit
1 options
Catalog No. MAVM0960R Supplier MilliporeSigma™ Supplier No. MAVM0960R
Includes
Base, Standard ring, Gasket pair, Support grid, Vacuum gauge, ON/OFF valve, Vacuum control, and External tubing
Filters to drain, standard multiwell plate, deep-well plate, or undersized plate
  • Has no internal parts subject to wear or damage
  • Automation-friendly—robotic arm can remove ring, access the plate, and then replace ring
Specifications
Includes HDPE Base, nylon standard ring with EPDM gaskets, support grid, FEP-lined Tygon™ tubing, valves and pressure gauge
Dimensions (L x W x H) 4.8 x 6 x 2.59 in. (12.2 x 15.2 x 6.6 cm)
Length (Metric) 12.2 cm
Width (Metric) 15.2 cm
Height (Metric) 6.6 cm