missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MultiScreen™HTS 96-Well Filter Plates for Clearing
Description
- Automation-compatible multiscreen filter plates
- Lysate clearing
Clearing of bacterial cell lysates
Specifications
Specifications
| Format | Standard |
| Membrane | Negatively Charged Phosphocellulose, Polyester Mesh |
| Array | 8 x 12 |
| Pore Size | 0.65 μm |
| Color | Clear |
| Material | Barex |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Filtration Area | 0.26 cm2 |
| For Use With (Application) | Protein kinase assays with peptide substrates, enzyme assays, binds positive charge |
| Show More |