missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MultiScreen™ Solvinert Filter Plates
Description
- Clear polyolefin base plate offers broad solvent compatibility
- One-piece design eliminates leaking and allows for extended incubations
- Low binding and low extractables
- Accommodate aqueous and nonaqueous samples with chemically resistant hydrophilic or hydrophobic membranes
- Polytetrafluoroethylene membranes feature 45μL pores
- Automation compatible
- Available in 96-well deep well (2mL) format (with or without pre-filter) and standard (500μL) format
Specifications
Specifications
| Format | Standard |
| Membrane | Hydrophobic PTFE |
| Array | 8 x 12 |
| Pore Size | 0.45 μm |
| Color | Clear |
| Material | Polyolefin Copolymer |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Filtration Area | 0.28 cm2 |
| For Use With (Application) | Solid phase synthesis and cleanup, in plate protein precipitation, sample recovery following in-plate compound cleavage, peptide synthesis |
| Show More |