Learn More
MilliporeSigma™ MultiScreen™ Multiple Punch and Punch Tips
Catalog No. MAMP09608
Change view
Click to view available options
Product Type:
Punch kit
Punch tip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
MAMP09608 Punch kit
MADP19650 Punch tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAMP09608 Supplier MilliporeSigma™ Supplier No. MAMP09608
For rdionucleotide counting by transfer of individual filters to tubes
  • Punch and tips are used to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioscope detection
  • Remove filters from wells separately, eliminating cross-contamination
  • Usable with all MultiScreen plates containing Durapore (PVDF) membrane; mixed cellulose ester membranes (HA) and glass fiber membranes must be wet when punched from plate
  • Multiple punch includes punch with punch carrier plate and punch distributor
Order Info
Disposable punch tips for the multiple punch are available. Three punch kits are available—all include multiple punch and carrier racks
Specifications
Product Type Punch kit
For Use With (Application) Radionucleotide counting by transfer of individual filters to tubes
Includes Punch with punch carrier plate and punch distributor
Quantity 1/Pk.