Learn More
MilliporeSigma™ MultiScreen™ Multiple Punch and Punch Tips
Description
- Punch and tips are used to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioscope detection
- Remove filters from wells separately, eliminating cross-contamination
- Usable with all MultiScreen plates containing Durapore (PVDF) membrane; mixed cellulose ester membranes (HA) and glass fiber membranes must be wet when punched from plate
- Multiple punch includes punch with punch carrier plate and punch distributor
Order Info
Disposable punch tips for the multiple punch are available. Three punch kits are available—all include multiple punch and carrier racks
Specifications
Specifications
| Product Type | Punch tip |
| For Use With (Application) | Radionucleotide counting by transfer of individual filters to tubes |
| Quantity | 50/Cs. |