missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MultiScreen™-Mesh Filter Plates with 96-Well Receiver Plates
Ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model
$1,105.90 - $1,375.38
Specifications
| Format | Standard |
|---|---|
| Membrane | Nylon Mesh |
| Array | 8 x 12 |
| Color | Clear |
| Material | Styrene |
| Catalog Number | Mfr. No. | Pore Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Pore Size | Price | Quantity & Availability | |||||
|
MANM10010
|
MilliporeSigma™
MANM10010 |
100 μm |
Pack of 10 Each for $1,190.00
|
|
|||||
|
MANMN2010
|
MilliporeSigma™
MANMN2010 |
20 μm |
Pack of 10 Each for $1,105.90
|
|
|||||
|
MANMN4010
|
MilliporeSigma™
MANMN4010 |
40 μm |
Pack of 10 Each for $1,349.75
|
|
|||||
|
MANMN6010
|
MilliporeSigma™
MANMN6010 |
60 μm |
Pack of 10 Each for $1,375.38
|
|
|||||
Description
96-well plate and 96-well receiver tray fit together to provide a complete system for culturing, testing, and reading assay results in a one-step protocol. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.
- Provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms
- Optically clear for use with an imaging microscope or other microscopic analysis
- Can be coated with a thin layer of agar if necessary
- Have also been shown to be useful as high-throughput cell strainers prior to cellular analysis in a FACS™ analyzer
Specifications
| Standard | |
| 8 x 12 | |
| Styrene | |
| Non-sterile | |
| 10/Pk. |
| Nylon Mesh | |
| Clear | |
| 96 | |
| Whole organism screening |