missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ High-Volume Manifold Collar

Catalog No. MSVMHTSHV
Change view
Click to view available options
Material:
Nylon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
MSVMHTSHV Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSVMHTSHV Supplier MilliporeSigma™ Supplier No. MSVMHTSHV

For use with MultiScreen high-volume filter plates

  • Fits over High-Volume Collection Plates
  • Opaque nylon construction

Specifications

Color Opaque
Material Nylon
For Use With (Application) Vacuum filtration