missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ Column Loaders

Catalog No. MACL09625
Change view
Click to view available options
Volume (Metric) Well:
25 μL
45 μL
100 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well
MACL09625 25 μL
MACL09600 100 μL
MACL09645 45 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MACL09625 Supplier MilliporeSigma™ Supplier No. MACL09625

No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly

  • Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating

Specifications

Format Standard
Array 8 x 12
Color Black
Volume (Metric) Well 25 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Load 25μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography
Quantity 1/Pk.