Learn More
MilliporeSigma™ MultiScreen™ Column Loaders
No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly
Supplier: MilliporeSigma™ MACL09600
Description
- Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating
Chemical Identifiers
| Black | |
| Non-sterile | |
| 1/Pk. |
Specifications
| Black | |
| 100 μL | |
| Load 100μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography |
| MultiScreen Column Loader | |
| Non-sterile | |
| 1/Pk. |