Learn More
MilliporeSigma™ MultiScreen™ Centrifuge Alignment Frame
Allows easy and accurate transfer of samples
Supplier:  MilliporeSigma™ MACF09604
Catalog No. MACF09604
Only null left
Explore available promotions
Description
Description
  • Transfer samples filtrate to the wells of a 96-well collection plate
  • Prevents spillage or cross-contamination
Specifications
Specifications
Blue
Non-sterile
4/Pk.
ABS Plastic
Use to align MultiScreen plate to 96-well receiver plate for centrifugation of aqueous media
Product Suggestions
Product Suggestions