missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay System Accessory: Feeder/Transport Trays

Catalog No. MACAC0RS5
Change view
Click to view available options
Quantity:
5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MACAC0RS5 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MACAC0RS5 Supplier MilliporeSigma™ Supplier No. MACAC0RS5

Includes

(5) 96-well Feeder Trays; (5) transport trays with lids

Perform cell growth, feeding, and analysis in one plate

  • With Lid

Specifications

Format Standard
Lid With Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Filtration Area 0.28 cm2
For Use With (Application) Caco-2 cell culture, cell-based assays
Quantity 5/Pk.