missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for Sample Preparation

Catalog No. MABVN1250
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
0.65 μm
1.2 μm
Color:
Clear
Material:
ABS Plastic
Styrene
Quantity:
10/Pk.
50/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Color Material Quantity
MABVN1250 1.2 μm Clear Styrene 50/Pk.
MADVN6550 0.65 μm Clear Styrene 50/Pk.
MAGVS2210 0.22 μm Clear ABS Plastic 10/Pk.
MAHVN4510 0.45 μm Clear Styrene 10/Pk.
MAHVN4550 0.45 μm Clear Styrene 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MABVN1250 Supplier MilliporeSigma™ Supplier No. MABVN1250

For the isolation, purification, and preparation of samples prior to testing in drug discovery processes and protein and DNA applications

Formats are suitable for removal of large particulates, resin-based separations, solvent applications, and protein/DNA concentration or purification. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold

Choice of filter materials

  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes

Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.

†MultiScreenHTS version features a recessed underdrain and rigid sidewalls.

Specifications

Format Standard
Membrane Hydrophilic PVDF
Array 8 x 12
Pore Size 1.2 μm
Color Clear
Product Type MultiScreen-BV Plate
Material Styrene
Volume (Metric) Well 0.05 to 0.25 mL
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28cm2
For Use With (Application) Receptor binding assays, compound generation sample prep, resin, bead based assays, sample prep
Quantity 50/Pk.
Show More Show Less