Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for ELISPOT Assays

Catalog No. MSIPS4W10
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Product Type:
MultiScreen HTS
MultiScreen HTS-IP with Underdrain
MultiScreen IP
MultiScreen-HA
MultiScreen-IP with Underdrain
Material:
Acrylic
Hydrophobic PVDF
Polystyrene
Membrane:
Immobilon-IP
Immobilon-P
Mixed Cellulose Esters
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material Membrane
MSIPS4W10 MultiScreen HTS Acrylic Immobilon-IP
MAHAS4510 MultiScreen-HA Polystyrene Mixed Cellulose Esters
MAIPS4510 MultiScreen-IP with Underdrain Acrylic Immobilon-P
MSIPN4W50 MultiScreen HTS-IP with Underdrain Acrylic Immobilon-IP
S2EM004M99 MultiScreen IP Acrylic Immobilon-P
351044 MultiScreen IP Hydrophobic PVDF Immobilon-P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
Catalog No. MSIPS4W10 missing translation for 'mfr' MilliporeSigma™ missing translation for 'supplierNo' MSIPS4W10
Only null left

Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining

  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days

Specifications

Format Standard
Lid Yes
Membrane Immobilon-IP
Array 8 x 12
Pore Size 0.45 μm
Product Line MultiScreen
Color White
Product Type MultiScreen HTS
Material Acrylic
No. of Wells 96
Sterility Sterile
Filtration Area 0.28 cm2
For Use With (Application) ELISPOT Assays
Quantity 10/Pk.
Show More Show Less