missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for Cell-Based Assays
Description
Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
Specifications
Specifications
| Format | Standard |
| Membrane | Polycarbonate |
| Array | 8 x 12 |
| Pore Size | 5 μm |
| Color | Clear |
| Material | Styrene |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Tissue Culture |
| Filtration Area | 0.28cm2 |
| Show More |