missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for Cell-Based Assays

Catalog No. MAMIC3S10
Change view
Click to view available options
Pore Size:
3 μm
5 μm
8 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
MAMIC3S10 3 μm
MAMIC5S10 5 μm
MAMIC8S10 8 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MAMIC3S10 Supplier MilliporeSigma™ Supplier No. MAMIC3S10

Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed

Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

Specifications

Format Standard
Membrane Polycarbonate
Array 8 x 12
Pore Size 3 μm
Color Clear
Material Styrene
No. of Wells 96
Sterility Sterile
Surface Treatment Tissue Culture
Filtration Area 0.28 cm2
For Use With (Application) Migration, invasion and chemotaxis
Quantity 10/Pk.
Show More Show Less