missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Monomethyl-Histone H4 (Lys20), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17651MI
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17651MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17651MI Supplier MilliporeSigma™ Supplier No. 17651

Rabbit Polyclonal Antibody

Specifically detects Monomethyl-Histone H4 (Lys20) Clone: in Human, Plant, Vertebrate samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Specifications

Antigen Monomethyl-Histone H4 (Lys20)
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-monomethyl-Histone H4 (Lys20) (rabbit polyclonal IgG). One vial containing 100μg of affinity purified antibody in 100μL 0.1M Tris-Glycine (pH 7.4), 15mM NaCl, and 0.05% NaN3. Normal Rabbit IgG. One vial containing 125μg of purified rabbit IgG in 125μL storage buffer. ChIP Primers GAPDH Coding Region. One vial containing 75μL of 5μM of each primer specific for human GAPDH coding region. FOR: GGC TCC CAC CTT TCT CAT CC; REV: GGC CAT CCA CAG TCT TCT GG
Gene Accession No. P62805
Gene Symbols H4/A , H4/B, H4/C , H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/J, H4/M, H4/N, H4/O , H4/a , H4F2, H4FA , H4FB , H4FC , H4FD, H4FE, H4FG, H4FH, H4FI , H4FJ, H4FK, H4FM , H4FN, H4FO, HIST2H4
Host Species Rabbit
Immunogen The immunogen was a synthetic peptide corresponding to amino acids 15-24 of histone H4, monomethylated on Lys20.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003538.3
Test Specificity Recognizes histone H4, Mr ∽11kDa.
Includes This ChIPAb+ Monomethyl-Histone H4 (Lys20) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Plant, Vertebrates
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only