missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Modified Barth's Saline (1X), liquid, without Ficoll™ 400

Catalog No. F04B
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04B Supplier MilliporeSigma™ Supplier No. F04B

Used in the culture of Xenopus Oocytes.

Specifications

Content And Storage 2-8°C up to 1 year
For Use With (Application) Culture of Xenopus Oocytes
Quantity 500 mL