Learn More
MilliporeSigma™ Millicell™ Culture Plate Inserts: HATF
Designed for improved cell differentiation and expanded applications involving analytical cell biology
$465.00 - $580.00
Specifications
Pore Size 0.45 μm
Sterility Sterile
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Filter Height (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Filter Height (Metric) Price Quantity  
PIHA01250
View Documents
MilliporeSigma™
PIHA01250
12mm 10.5mm
Pack of 50 Each for $465.00
Only null left
PIHA03050
missing translation for 'viewDocuments'
MilliporeSigma™
PIHA03050
30mm 13mm
Pack of 50 Each for $580.00
Only null left
Description
Description
  • Microporous membranes allow access to cell monolayer from both apical and basolateral domains; promote enhanced cell attachment, growth, and differentiation of attachment-dependent and suspension cultures
  • Ready-to-use Milicell inserts fit most 6- or 24-well plates and are easily prepared for SEM and TEM techniques; compatible with standard and fluorescent stains
  • Effective growth area for 12mm diameter inserts, 0.6cm2; and 4.2cm2 for 30mm diameter inserts
  • Don't require an extracellular matrix for attachment-dependent cells
  • Polystyrene housing; supplied sterile and individually blister packed
  • Inserts utilize a mixed cellulose esters membrane; Triton™-free
Applications:
Transport and permeability studies, co-culture for cell-cell interaction studies, in vitro toxicity testing, tumor cell metastasis studies, and organotypic cultures
Specifications
Specifications
0.45 μm
Sterile