missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ pH Test Strips and Indicator Papers
Measure pH onsite without instruments or extensive preparation
$95.70 - $620.75
Specifications
| For Use With (Application) | Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination |
|---|---|
| Storage Requirements | 10°C to 25°C |
| Catalog Number | Mfr. No. | pH Range | Type | Quantity | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | pH Range | Type | Quantity | Price | Quantity & Availability | |||
|
M1095330001
|
MilliporeSigma™
1.09533.0001 |
5 to 10 | Neutralit pH Indicator Strip | 100 Strips |
Each for $184.51
|
|
|||
|
M1095310001
|
MilliporeSigma™
1.09531.0001 |
0 to 6 | Acilit pH Indicator Strip | 100 Strips/Ea. |
Each for $184.51
|
|
|||
|
M1095320001
|
MilliporeSigma™
1.09532.0001 |
7.5 to 14 | Acilit pH Indicator Strip | 100 Strips in Plastic Box |
Each for $187.88
|
|
|||
|
M1095400001
|
MilliporeSigma™
1.09540.0001 |
0 to 2.5 | Special pH Indicator Strip | 100 Box |
Each for $179.28
|
|
|||
|
M1095350001
|
MilliporeSigma™
1.09535.0001 |
0 to 14 | Universal pH Indicator Strip | 100 Strips |
Each for $175.25
|
|
|||
|
M1095410001
|
MilliporeSigma™
1.09541.0001 |
2.5 to 4.5 | Special pH Indicator Strip | 1 Box |
Each for $123.88
|
|
|||
|
M1095430001
|
MilliporeSigma™
1.09543.0001 |
6.5 to 10 | Special pH Indicator Strip | 1 Box of 100 |
Each for $182.33
|
|
|||
|
M1095420001
|
MilliporeSigma™
1.09542.0001 |
4 to 7 | Special pH Indicator Strip | 100 Box |
Each for $126.96
|
|
|||
|
M1095620003
|
MilliporeSigma™
1.09562.0003 |
9.5 to 13 | Acilit pH Indicator Paper | 3 x 4.8 Rolls |
Each for $132.51
|
|
|||
|
M1095450001
|
MilliporeSigma™
1.09545.0001 |
11 to 13 | Special pH Indicator Strip | 1 Box |
Each for $95.70
|
|
|||
|
M1095840001
|
MilliporeSigma™
1.09584.0001 |
2 to 9 | Standard pH Indicator Strip | 100 Test Strips |
Each for $179.28
|
|
|||
|
M1095330007
|
MilliporeSigma™
1.09533.0007 |
5 to 10 | Neutralit pH Indicator Strip | 6 Pack |
Pack of 6 Each for $620.75
|
|
|||
|
M1095310007
|
MilliporeSigma™
1.09531.0007 |
0 to 6 | Acilit pH Indicator Strip | 6 Pack |
Pack of 6 Each for $444.27
|
|
|||
|
M1095350007
|
MilliporeSigma™
1.09535.0007 |
0 to 14 | Universal pH Indicator Strip | 6 Pack |
Pack of 6 Each for $556.99
|
|
|||
|
M1095420007
|
MilliporeSigma™
1.09542.0007 |
4 to 7 | Special pH Indicator Strip | 6 Box |
Pack of 6 Each for $601.64
|
|
|||
|
M1095430007
|
MilliporeSigma™
1.09543.0007 |
6.5 to 10 | Special pH Indicator Strip | 6 Box |
Pack of 6 Each for $459.16
|
|
|||
|
M1095600003
|
MilliporeSigma™
1.09560.0003 |
0.5 to 5 | Acilit pH Indicator Paper | 3 x 4.8 Rolls |
Each for $139.28
|
|
|||
Description
Fit into a pocket and provide easy, rapid results—suited to all media in environmental analysis and industrial in-process controls.
- Differentiated color gradations for a more precise evaluation
- Traditional indicator paper or MQuant™ nonbleeding universal strips available
- High-quality papers impregnated with indicator or mixed-indicator solutions, cut to size and dried
- Supplied in a roll format that protects strips against moisture, light or ambient gases for longer storage life
- Special indicator dyes covalently bonded to reagent papers
- Indicator does not bleed, so strips will not contaminate the medium, even if left for long periods of time
- Can be used for three to five years when stored at 10 to 25°C in a light-protected area
- Reseal quickly after each use
- Tests checked and calibrated using certified buffer solutions
- Buffer solutions are traceable to primary reference materials from NIST™ and PTB
Order Info
Packaging: indicator papers come in lengths of 4.8m in roll dispenser; MQuant™ special indicator pH strips come as strips packed in boxes of 100 tests each.
Specifications
| Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer Solutions, rapid on-the-spot pH determination, non-bleeding even in strong alkaline solutions for accurate pH determination | |
| 10°C to 25°C |
Safety and Handling
ShelfLife : 3 to 5 Years