missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Luminata™ Classico Western HRP substrate, 100mL

Catalog No. WBLUC0100A
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WBLUC0100A 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WBLUC0100A Supplier MilliporeSigma™ Supplier No. WBLUC0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Premixed, ready to use chemiluminescent HRP detection reagent

  • The reagent has been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays.

  • Premixed for convenience to avoid cross contamination of the reagents.
  • Broad range of sensitivities to satisfy all detection needs.
  • Consistent results with less pipetting error.
  • Stable at 4°C and also at room temperature which allows storage on bench.
  • Best detection performance relative to competition.
  • Compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Western blotting, dot/slot blotting

Specifications

Storage Requirements 2° to 8°C
For Use With (Application) Dot/Slot Blot, Western Blot
Quantity 100 mL