missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Low Trypsin-High EDTA

Catalog No. SM2004C
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SM2004C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SM2004C Supplier MilliporeSigma™ Supplier No. SM2004C

For cell culture applications

  • PBS Based
  • Contains 0.025% Trypsin and 0.75mM EDTA (1X)
  • Formulated without Ca2+ and Mg2+

Specifications

Content And Storage -20° C for 18 months
Form Liquid
Product Type Cell Culture Dissociation Reagent
For Use With (Application) Cell Culture
Quantity 100 mL