Learn More
MilliporeSigma™ Integrity Test Kit for Small Volume Devices
Catalog No. SLTEST000
Change view
Click to view available options
Pore Size:
5 μm
Catalog No. Pore Size
SLTEST000 5 μm
1 options
Catalog No. SLTEST000 Supplier MilliporeSigma™ Supplier No. SLTEST000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For small volume devices
  • Convenient bubble-point testing
  • For vented and non-vented devices
  • Tests wide range of pore sizes and membrane types
  • Millipore membrane filter units can be integrity-tested using the bubble point test method. Small, syringe-driven membrane filter units with 15 cm2 filtration area, such as Millex and Sterivex units, can be conveniently tested using the syringe-style integrity tester.
    Specifications
    Pore Size 5 μm
    Filtration Area 15 cm2
    Type Integrity Tester