missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immobilon™ Western Chemiluminescent HRP Substrate (ECL)

Catalog No. WBKLS0050
Change view
Click to view available options
Quantity:
2 x 25 mL
2 x 50 mL
2 x 250 mL
Sufficient For:
1000cm2 Membrane
5000cm2 Membrane
500cm2 Membrane
Kit Contents:
250mL luminol, 250mL peroxide solution
25mL luminol, 25mL peroxide solution
50mL luminol, 50mL peroxide solution
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For Kit Contents
WBKLS0050 2 x 25 mL 500cm2 Membrane 25mL luminol, 25mL peroxide solution
WBKLS0500 2 x 250 mL 5000cm2 Membrane 250mL luminol, 250mL peroxide solution
WBKLS0100 2 x 50 mL 1000cm2 Membrane 50mL luminol, 50mL peroxide solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. WBKLS0050 Supplier MilliporeSigma™ Supplier No. WBKLS0050

Includes

Luminol, peroxide solution

Provides high sensitivity over a broad detection range at an economical price

  • Combines high sensitivity and low background
  • Allows detection of high- and low-abundance proteins in the mid-femtogram range
  • Lowers antibody consumption by two- to five-fold compared to less sensitive reagents
  • Compatible with PVDF and nitrocellulose membranes as well as all commonly used buffers and blocking reagents

Specifications

For Use With (Application) Dot/Slot Blot
Quantity 2 x 25 mL
Substrate Horseradish Peroxidase
Sufficient For 500cm2 Membrane
Kit Contents 25mL luminol, 25mL peroxide solution
Content And Storage 2°C to 8°C