missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Human VEGF165 Recombinant Protein, Animal Free
Specifications
Specifications
| Name | Human VEGF165 Recombinant Protein, Animal Free |
| Quantity | 10 μg |
| Source | E. coli |
| Cross Reactivity | Mouse, Rat, Pig, Hamster, Trout, Green Algae, Sheep, Cow, Rabbit, Guinea Pig |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin free and greater than 98% pure by SDS-PAGE gel and HPLC analyses |