missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human VEGF121 Recombinant Protein, Animal Free

Catalog No. GF349
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF349 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF349 Supplier MilliporeSigma™ Supplier No. GF349

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Name Human VEGF121 Recombinant Protein
Quantity 10 μg
Source E. coli
Recombinant Recombinant
Purity or Quality Grade Endotoxin level is less than 0.1ng/μg of protein