missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human TGF-β1 Recombinant Protein, Animal Free

Supplier:  MilliporeSigma™ GF346

Catalog No. GF346


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Human TGF-beta1 Recombinant Protein
E. coli
Endotoxin level is less than 0.1ng/μg of protein
10 μg
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions