missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Human LIF Recombinant Protein, Animal Free
Specifications
Specifications
| Name | Human LIF Recombinant Protein |
| Quantity | 25 μg |
| Source | E. coli |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin level is less than 0.1ng/μg of protein |