missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Human IL-22 Recombinant Protein, Animal Free
Specifications
Specifications
| Name | Human IL-22 Recombinant Protein |
| Quantity | 10 μg |
| Source | E. coli |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin level is less than 0.1ng/μg of protein |