missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ HDAC3 Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710238MI
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
1710238MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 1710238MI missing translation for 'mfr' MilliporeSigma™ missing translation for 'supplierNo' 1710238

Mouse Monoclonal Antibody

Specifically detects HDAC3 Clone: in Bovine, Hamster, Human, Mouse samples, and it is validated for ChIP, Western Blotting

Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. HDAC3 belongs to the histone deacetylase/acuc/apha family. It has histone deacetylase activity and represses transcription when tethered to a promoter. It may participate in the regulation of transcription through its binding with the zinc-finger transcription factor YY1. This protein can also down-regulate p53 function and thus modulate cell growth and apoptosis. HDAC3 is regarded as a potential tumor suppressor gene.

Specifications

Antigen HDAC3
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-HDAC3 (mouse monoclonal). One vial containing 100μg of protein G purified monoclonal in buffer containing 0.02M phosphate buffer (pH 7.6), 0.25M NaCl, 0.1% sodium azide and 30% glycerol. Concentration: 1mg/mL. Normal Mouse IgG. One vial containing 125μg of purified mouse IgG in 125μL of storage buffer containing 0.1% sodium azide. ChIP Primers, p21. One vial containing 75μL of each primer (5μM) specific for a region of the human p21 (WAF1/CIP1/CDKN1A) promoter. FOR: CCC ACA GCA GAG GAG AAA GAA; REV: CTG GAA ATC TCT GCC CAG ACA
Gene Accession No. O15379
Gene Symbols HDAC3, SMAP45, HD3, RPD3, RPD3-2
Host Species Mouse
Immunogen peptide (NEFYDGDHDNDKESDVEI) corresponding to amino acids 411-428 of human HDAC3
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_003883
Includes This ChIPAb+ HDAC3 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Bovine, Hamster, Human, Mouse
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only