missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Hazardous Waste Filtration System Support Screen
Replacement part for Hazardous Waste Filtration System
Supplier: MilliporeSigma™ YT4014259
Description
- Stainless-Steel construction
- Diameter: 142mm
- PTFE-coated
Specifications
| Hazardous Waste Filtration System | |
| PTFE-coated Stainless Steel | |
| Support Screen | |
| 142 mm |