missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ GS System 100mM L-Methionine sulfoximine
Description
The GS System™ (offered by and a registered trademark of Celltech Biologicals, Inc.) utilizes specific vectors for cell culture transfection. Specialty Media offers media and reagents for use with this system. Our GS System™ products are made using the highest quality reagents and our high purity Ultra Pure H
Specifications
Specifications
| Content And Storage | -20°C up to 1 year |
| For Use With (Application) | Utilizes specific vectors for cell culture transfection |
| Quantity | 10 mL |