Learn More
MilliporeSigma™ Accessories for Fluid Sampling Kit
Catalog No. XX6403780
Change view
Click to view available options
Type:
Remote Sampling Assembly
Sampler Valve Hose Assembly
Tubing
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
XX6403780 Tubing
XX6403705 Remote Sampling Assembly
XX6403708 Sampler Valve Hose Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. XX6403780 Supplier MilliporeSigma™ Supplier No. XX6403780
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessories for filter holders for sample preparation.
Fluid Sampling Kit Components and Accessories
Specifications
Autoclavable Autoclavable
Material Tygon™
For Use With (Application) Sampling turbine fuels, water or other liquids through Millipore™ contamination analysis monitors prior to gravimetric or colorimetric analysis
Type Tubing
Diameter (English) Inside 0.18 in.
Diameter (Metric) Inside 0.45 cm
For Use With (Equipment) Fluid Sampling Kit