Learn More
MilliporeSigma™ Fluid Sampling Kit
Accessories for filter holders for sample preparation.
Supplier: MilliporeSigma™ XX6403730
Description
The Fluid Sampling Kit is a complete solution for field-based fluid collection and contamination analysis. The stainless steel sampler assembly plugs into a quick-release valve installed in the system line, which allows a measured quantity of liquid to pass through the monitor. A valved syringe serves as a pump to remove residual liquid from the monitor. Kit is compatible with our 37 mm contamination monitors and matched-weight monitors.For gravimetric analysis, accessory Matched-Weight Monitors eliminate filter pre-weighing before sampling.Features & Benefits:•Sturdy carrying case and stainless steel sampling assembly provide years of in-field service •Fluid Sampling Kit includes 12 Tenite™ contamination monitors Applications:For sampling turbine fuels, water or other liquids through Millipore Contamination Analysis Monitors prior to gravimetric or colorimetric analysis.Specifications
| Stainless Steel with Stainless-Clad PTFE Hose; Polystyrene Monitors | |
| Quick release valve or plug, 1/8 in. NPTF thread, Viton A seals | |
| 9.25 in. | |
| 4.88 in. |
| Sampling turbine fuels, water or other liquids through Millipore™ contamination analysis monitors prior to gravimetric or colorimetric analysis | |
| Fluid Sampling Kit | |
| 23.5 cm | |
| 12.4 cm |