Learn More
MilliporeSigma™ Flow-limiting Orifices
Creates quantitative airflow through a filter for airborne contamination analysis
Specifications
| Product Type | Environmental Test Kits or Supplies |
|---|---|
| Material | Stainless Steel |
| For Use With (Equipment) | Millipore Aerosol Analysis Holder or Aerosol Adapter |
| For Use With (Application) | To create quantitative airflow through a Millipore filter for airborne contamination analysis |
Description
Flow Limiting Orifices create quantitative airflow through a MF-Millipore filter for airborne contamination analysis. The orifices are threaded into the outlet of a Millipore Aerosol Analysis Holder or Aerosol Adapter to enable a constant flow rate when required vacuum level is applied.Ratings are accurate to within approximately 15%. For greater precision, calibrate orifice with a flowmeter at actual working vacuum level.Specifications
| Environmental Test Kits or Supplies | |
| Stainless Steel | |
| Millipore Aerosol Analysis Holder or Aerosol Adapter | |
| To create quantitative airflow through a Millipore filter for airborne contamination analysis |