Learn More
MilliporeSigma™ EZ-Fit™ Manifolds
General microbiological analysis
Supplier: MilliporeSigma™ EZFITMIC06
Description
EZ-Fit™ Manifold is available with the new Microfil™ head for use with presterilized, ready-to-use funnels and membranes.Specifications
| Autoclavable | |
| Microfil funnels and membranes | |
| Stainless Steel | |
| 6-places |