missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ESGRO Complete™ Gelatin Solution

Catalog No. SF008
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF008 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF008 Supplier MilliporeSigma™ Supplier No. SF008

For stem cell culture

Specifications

Form Gelatin Solution
For Use With (Application) Stem Cell Culture
Quantity 500 mL