missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ESGRO Complete™ Accutase™ Cell Detachment Solution

Catalog No. SF006MI
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF006MI 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF006MI Supplier MilliporeSigma™ Supplier No. SF006

a cell detachment solution of proteolytic and collagenolytic enzymes, qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete Clonal Grade Medium.

ESGRO Complete™ Accutase™ is a cell detachment solution of proteolytic and collagenolytic enzymes that has been qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete™ Clonal Grade Medium (Cat. No. SF001-500). Accutase™ does not contain mammalian or bacterial derived products.

Specifications

Content And Storage Stable when stored at -20° C
For Use With (Application) Stem Cell Culture
Quantity 100 mL