missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ESGRO™ Chemicon™ 2i Supplement Kit (1000x)

Catalog No. ESG1121
Change view
Click to view available options
Quantity:
1 Kit/5 L medium
1 Kit/500 mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ESG1121 1 Kit/500 mL medium
ESG1120 1 Kit/5 L medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ESG1121 Supplier MilliporeSigma™ Supplier No. ESG1121

contains LIF a selective GSK3β and Mek 1/2 inhibitors to enhance viability of mouse embryonic stem and iPS cells in the absence of serum and feeder cells

The ESGRO™-2i Supplement Kit (1000x) contains LIF a selective GSK3β and Mek 1/2 inhibitors to enhance viability of mouse embryonic stem and iPS cells in the absence of serum and feeder cells. Supports a naive/ground pluripotent state.

Specifications

Content And Storage ESGRO Supplement: (Mfr. No. CS210511): Mouse recombinant Leukemia Inhibitory Factor (LIF), 0.5 x 106U/mL. One (1) 0.5mL vial. MEK/GS3 Inhibitor Supplement: (Mfr.. No. CS210510-500UL): One (1) 0.5mL vial. Store at -20°C up to 2 years
For Use With (Application) Stem cell culture
Quantity 1 Kit/500 mL medium