missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Enzyme Free Cell Dissociation Solution PBS Based (1x)

Catalog No. S014C
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S014C 100 mL
S014B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S014C Supplier MilliporeSigma™ Supplier No. S014C

Used to dissociate primary cells, tissues, and tumors with an increased plating efficiency. MilliporeSigma™ Enzyme Free Cell Dissociation Solution PBS Based (1x) is PBS based formulation of chelating agents that contain agents to stabilize their activity on cells.

  • Works significantly faster than 0.5mM EDTA
  • Preserves structural and functional integrity of cell surface proteins
  • Does not have cytotoxic effects sometimes associated with 0.5 mM EDTA
  • Used with cells that are sub-confluent (60-80%)

Specifications

Content And Storage Store at room temperature or at 4°C.
Concentration 1 X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Formulation Chelating and stabilizing agents
For Use With (Application) Cell Dissociation
Quantity 100 mL